missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adjustment Bar

OHAUS™ Adjustment Bar is an accessory for use with adjustable platforms, which are designed to be mounted onto heavy duty shakers.

66.65 € - 82.00 €

Specifications

Product Type Adjustment Bar
Color Black
For Use With (Equipment) Heavy Duty Shakers
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
15752489
View Documents
OHAUS™
30400073
460 mm
66.65 €
Each
In Stock
to see stock.
Add to basket
 
15762489
View Documents
OHAUS™
30400074
610 mm
82.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • For use with platforms, which increase the amount of usable space for samples
  • Heavy duty shaker accessory
Specifications

Specifications

Adjustment Bar
Black
Heavy Duty Shakers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.