Learn More
Adipoyl chloride, 98%, Thermo Scientific Chemicals
46.73 € - 572.85 €
Chemical Identifiers
CAS | 111-50-2 |
---|---|
Molecular Formula | C6H8Cl2O2 |
Molecular Weight (g/mol) | 183.028 |
MDL Number | MFCD00000759 |
InChI Key | PWAXUOGZOSVGBO-UHFFFAOYSA-N |
Synonym | adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242 |
PubChem CID | 61034 |
IUPAC Name | hexanedioyl dichloride |
SMILES | C(CCC(=O)Cl)CC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11454554
|
Thermo Scientific Alfa Aesar
A13168.14 |
25 g |
46.73 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464554
|
Thermo Scientific Alfa Aesar
A13168.22 |
100 g |
129.65 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474554
|
Thermo Scientific Alfa Aesar
A13168.36 |
500 g |
572.85 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.
Solubility
Immiscible with water.
Notes
Incompatible strong oxidizing agents, bases and acids.
Chemical Identifiers
111-50-2 | |
183.028 | |
PWAXUOGZOSVGBO-UHFFFAOYSA-N | |
61034 | |
C(CCC(=O)Cl)CC(=O)Cl |
C6H8Cl2O2 | |
MFCD00000759 | |
adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242 | |
hexanedioyl dichloride |
Specifications
111-50-2 | |
105°C to 107°C (2 mmHg) | |
Acrid | |
ClCO(CH2)4COCl | |
MFCD00000759 | |
507709 | |
adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242 | |
PWAXUOGZOSVGBO-UHFFFAOYSA-N | |
hexanedioyl dichloride | |
61034 | |
98% |
1.255 | |
>110°C (230°F) | |
C6H8Cl2O2 | |
1.4705 | |
UN3265 | |
Moisture sensitive | |
Immiscible with water. | |
C(CCC(=O)Cl)CC(=O)Cl | |
183.028 | |
183.03 | |
Adipoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 203-876-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Adipoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Adipoyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.