missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Additional Tips for Stainless Steel Samplers

Interchangeable tips to tailor your stainless steel samplers to your exact needs.

Brand:  Sampling Systems™ TIP-40P

 View more versions of this product

Product Code. 15849741

  • 326.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pointed tips used with hard, dense powders
  • Rounded tips used when you need to minimize the risk of damaging your equipment
  • Available to suit diameters of 25 and 40mm
  • Manufactured from 316L stainless steel
Specifications

Specifications

Interchangeable Sampler Tips
40 mm
Samplers
Stainless Steel (316L)
Pointed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.