missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Additional Tips for Stainless Steel Samplers
Interchangeable tips to tailor your stainless steel samplers to your exact needs.
Brand: Sampling Systems™ TIP-25R
This item is not returnable.
View return policy
Description
- Pointed tips used with hard, dense powders
- Rounded tips used when you need to minimize the risk of damaging your equipment
- Available to suit diameters of 25 and 40mm
- Manufactured from 316L stainless steel
Specifications
Interchangeable Sampler Tips | |
25 mm | |
Samplers |
Stainless Steel (316L) | |
Rounded |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction