Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For S450
Ideal for ramp operation with rapid temperature changes during incubation, breeding or storage.
Brand:  Memmert™ S450E20182
Product Code. 16305867
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with compressor-cooled incubator models 450 and 750
  • Grid pattern allows for airflow
  • Manufactured from electro-polished stainless steel
Specifications
Specifications
Models 450 and 750 Ovens
Grid
Stainless Steel