Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand: Eppendorf™ 5910712006
This item is not returnable.
View return policy
Description
- Suitable for S-4x400 Rotor buckets
- Accommodates selected array of tubes or bottles
Specifications
Skirted/Self-standing | |
50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge | |
S-4x400 Bucket | |
2/Pk. |