Learn More
Eppendorf™ Adapter S-4x Universal Rotor Large
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4xUniversal-Large rotor.
Brand: Eppendorf™ 5920751000
This item is not returnable.
View return policy
Description
- Suitable for S-4xUniversal-Large rotor buckets
- Accommodates selected array of tubes, bottles, or cards
Specifications
Round Bottom | |
17.5 mm | |
S-4xUniversal-Large | |
24 |
24 x 14 mL Round Bottom Tubes | |
123 mm | |
24 | |
2/Pk. |