missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25900-1-AP-20UL
This item is not returnable.
View return policy
Description
Proinflammatory cytokine tumor necrosis factor-a (TNF-a) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-a, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-a, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM 10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.Specifications
ADAM10 | |
Polyclonal | |
Unconjugated | |
ADAM10 | |
AD10, ADAM 10, ADAM10, CD156c, CDw156, HsT18717, kuz, Kuzbanian protein homolog, MADM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102, 11487 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14672, O35598 | |
ADAM10 | |
ADAM10 Fusion Protein Ag23078 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction