Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Classic and universal, these micropipettes are intended for all applications and come in a select three-pack. Socorex™ Acura™ Manual 825 Micropipette Triopack micropipettes are designed for comfort and sturdiness.
Brand: Socorex™ 825.TRIO.T
This item is not returnable.
View return policy
Description
- Ergonomic shape.
- Lightweight.
- Uultra-soft plunger stroke.
- Smart and reliable volume adjustment:
- State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
- Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
- Digital display of set volume on the front of the instrument is easily visible during pipetting.
- Autoclavable.
Specifications
EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices | |
Acura | |
5 to 50, 20 to 200, 100 to 1000 μL | |
3/Pk. |
Autoclavable | |
Micropipette Triopack | |
3 Acura™ micropipettes, manual, spike samples, Qualitips™ 50/200/1000μL |