missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13537-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene, and related gene, ACTL7A, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7B gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. Unlike ACTL7A, the ACTL7B gene is expressed predomitly in the testis, however, its exact function is not known.Specifications
ACTL7B | |
Polyclonal | |
Unconjugated | |
ACTL7B | |
Actin like 7 beta, actin like 7B, Actin like protein 7B, ACTL7B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10880, 11471, 313183 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4QR76, Q9QY83, Q9Y614 | |
Actl7b | |
ACTL7B Fusion Protein Ag4460 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |