missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, purum, Honeywell Fluka™
Purum, powder
78.72 € - 488.46 €
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.011 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15650790
|
Honeywell-Fluka
05120-250G |
250 g | |||||||
15640790
|
Honeywell-Fluka
05120-1KG |
1 kg |
148.73 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15660790
|
Honeywell-Fluka
05120-5KG |
5 kg |
488.46 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7440-44-0 | |
12.011 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
C | |
NONH for all modes of transport | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12.01g/mol | |
Activated Charcoal |
≤7.0% | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.011 | |
CHEBI:27594 | |
purum |
Safety and Handling
missing translation for 'einecsNumber' : 231-153-3