missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT(R) SA 2, Thermo Scientific Chemicals

Product Code. 10185293
Click to view available options
Quantity:
250 g
2.5 kg
25 kg
Packaging:
Fibre drum
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3652.0°C
Color Black
Loss on Drying 10% max.
Packaging Fibre drum
Quantity 25 kg
Merck Index 15, 1807
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
Formula Weight 12
Physical Form Powder
Chemical Name or Material Activated charcoal,NORIT* SA 2, NORIT™ SA 2, Decolorizing
Product Identifier
  • Activated charcoal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.