missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13989-1-AP-20UL
This item is not returnable.
View return policy
Description
An anti-rat ACSL1 antibody recognized a band of the predicted 68 kDa in high-speed supernatant from rat liver and in human and murine SMCs, monocyte-derived macrophages, and murine peritoneal macrophages. It has 2 isoforms produced by alternative splicing
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.Specifications
ACSL1 | |
Polyclonal | |
Unconjugated | |
ACSL1 | |
ACS1, ACSL1, Acyl CoA synthetase 1, FACL1, FACL2, LACS, LACS 1, LACS 2, LACS1, LACS2, Palmitoyl CoA ligase 1, Palmitoyl CoA ligase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14081, 2180, 25288 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18163, P33121, P41216 | |
ACSL1 | |
ACSL1 Fusion Protein Ag5059 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction