Learn More
BioAcryl-P (40%, 29:1), Thermo Scientific Chemicals
BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.
116.58 € - 191.96 €
Specifications
Chemical Name or Material | Acrylamide/Bisacrylamide 29:1 |
---|---|
Physical Form | Liquid |
Concentration | 40% soln |
Color | Colorless |
MDL Number | MFCD00080848 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455349
|
Thermo Scientific Alfa Aesar
J63079.AP |
500 mL |
116.58 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465349
|
Thermo Scientific Alfa Aesar
J63079.K2 |
1 L |
191.96 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
- Acrylamide is a crystalline amide that forms polymer structures at room temperature
- Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
- Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
Specifications
Acrylamide/Bisacrylamide 29:1 | |
40% soln | |
MFCD00080848 | |
It is soluble in water. |
Liquid | |
Colorless | |
UN3426 |
Safety and Handling
GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H340-H350-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.