Learn More
BioAcryl-P (30%, 29:1), Thermo Scientific Chemicals
BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.
134.67 € - 217.08 €
Specifications
Chemical Name or Material | Acrylamide/Bisacrylamide 29:1 |
---|---|
Physical Form | Liquid |
Concentration | 30% soln |
Color | Colorless |
MDL Number | MFCD00080848 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465549
|
Thermo Scientific Alfa Aesar
J63279.AP |
500 mL |
134.67 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475549
|
Thermo Scientific Alfa Aesar
J63279.K2 |
1 L |
217.08 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
• Acrylamide is a crystalline amide that forms polymer structures at room temperature
• Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
• Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
Specifications
Acrylamide/Bisacrylamide 29:1 | |
30% soln | |
MFCD00080848 | |
Difficult to mix. |
Liquid | |
Colorless | |
UN3426 |
Safety and Handling
GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302+H332-H315-H317-H319-H340-H350-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- BioAcryl-P (30%, 29:1)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.