missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetone, Reagent Grade, ≥99%, Honeywell™

Product Code. 15658050
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Color Colorless
Density 0.975g/mL at 25°C
pH 6 (at 200.00g/L)
Boiling Point 139.5°C (1013 hPa)
Flash Point 38°C (100.4°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3COCH2COCH3
Quantity 2.5 L
UN Number UN2310
Beilstein 741937
Solubility Information 153.8 - 155.2g/L at 20°C in water
Vapor Density 3.5 (vs air)
Formula Weight 100.12g/mol
Percent Purity ≥99%
Chemical Name or Material Acetylacetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetylacetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311 + H331-Toxic in contact with skin or if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.