missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylacetone, Reagent Grade, ≥99%, Honeywell™
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -23°C |
Color | Colorless |
Density | 0.975g/mL at 25°C |
pH | 6 (at 200.00g/L) |
Boiling Point | 139.5°C (1013 hPa) |
Flash Point | 38°C (100.4°F) |
Odor | Characteristic |
Packaging | Glass bottle |
Linear Formula | CH3COCH2COCH3 |
Quantity | 1 L |
Show More |
Safety and Handling
Product Identifier
- Acetylacetone
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Acute toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311 + H331-Toxic in contact with skin or if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction