Learn More
Acetovanillone, 98%, Thermo Scientific Chemicals
35.90 € - 216.00 €
Chemical Identifiers
CAS | 498-02-2 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00008747 |
InChI Key | DFYRUELUNQRZTB-UHFFFAOYSA-N |
Synonym | acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon |
PubChem CID | 2214 |
ChEBI | CHEBI:2781 |
IUPAC Name | 1-(4-hydroxy-3-methoxyphenyl)ethanone |
SMILES | COC1=CC(=CC=C1O)C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10070540
|
Thermo Scientific Acros
102420250 |
25 g | Glass bottle |
35.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10152940
|
Thermo Scientific Acros
102421000 |
100 g | Glass bottle |
77.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10723465
|
Thermo Scientific Acros
102425000 |
500 g | Glass bottle |
216.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
498-02-2 | |
166.18 | |
DFYRUELUNQRZTB-UHFFFAOYSA-N | |
2214 | |
1-(4-hydroxy-3-methoxyphenyl)ethanone |
C9H10O3 | |
MFCD00008747 | |
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
CHEBI:2781 | |
COC1=CC(=CC=C1O)C(C)=O |
Specifications
498-02-2 | |
100.0 | |
Yellow to Brown | |
Authentic | |
C9H10O3 | |
MFCD00008747 | |
15, 728 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in ethanol,acetone,benzene,very soluble in diethyl ether,chloroform,practically insoluble in petroleum ether | |
COC1=CC(=CC=C1O)C(C)=O | |
166.18 | |
CHEBI:2781 | |
98% | |
Acetovanillone |
97.5 | |
112.0°C to 116.0°C | |
295.0°C to 300.0°C | |
98% | |
HOC6H3(OCH3)COCH3 | |
08, 272 | |
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
DFYRUELUNQRZTB-UHFFFAOYSA-N | |
1-(4-hydroxy-3-methoxyphenyl)ethanone | |
2214 | |
166.18 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-854-5
RUO – Research Use Only