missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, puriss. p.a. ACS Reagent, Honeywell
150.78 € - 3322.99 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15683870
|
Honeywell Riedel-de Haën
33019-1L |
1 L |
150.78 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15693870
|
Honeywell Riedel-de Haën
33019-2.5L |
2.5 L |
321.18 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603880
|
Honeywell Riedel-de Haën
33019-25L |
25 L |
3322.99 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
75-05-8 | |
0.16 | |
973°F | |
CH3CN | |
72.8mmHg (20°C) | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
ACS/puriss. p.a. |
0.786g/mL at 25°C | |
2°C (35.6°F) | |
C2H3N | |
MFCD00001878 | |
1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.5% (GC) | |
Acetonitrile, Reag. Ph. Eur. |
Safety and Handling
P210-P280-P305 + P351 + P338
H225-H302 + H312 + H332-H319
Danger
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000