Learn More
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Solvent for LC-MS analysis
278.39 € - 1620.00 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492857
|
Thermo Scientific Alfa Aesar
047138.K2 |
1 L |
278.39 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412867
|
Thermo Scientific Alfa Aesar
047138.M1 |
4 x 1 L |
878.37 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402867
|
Thermo Scientific Alfa Aesar
047138.K7 |
4 L | N/A | ||||||
15422867
|
Thermo Scientific Alfa Aesar
047138.M6 |
4 x 4 L | N/A | ||||||
Description
Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
-48°C to -45°C | |
Colorless | |
80°C to 82°C | |
Pungent | |
1.344 | |
MFCD00001878 | |
741857 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
LC-MS |
75-05-8 | |
0.786 g/mL | |
5°C (41°F) | |
Liquid | |
C2H3N | |
UN1648 | |
14,70 | |
Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons. | |
CC#N | |
41.053 | |
CHEBI:38472 | |
99.8% | |
Acetonitrile |
Safety and Handling
GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ACETONITRILE
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetonitrile
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.