missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, HPLC for Fluorescence Detection, Fisher Chemical™
153.00 € - 272.00 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Weight (g/mol) | 41.05 |
MDL Number | 1878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10040010
|
Fisher Chemical
A/0630/PB15 |
1 L |
153.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10325120
|
Fisher Chemical
A/0630/PB17 |
2.5 L |
272.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
75-05-8 | |
1878 | |
acetonitrile |
41.05 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CC#N |
Specifications
-46°C | |
Colorless | |
Plastic-coated Glass Bottle | |
C2H3N | |
97mbar at 20°C | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
41.05g/mol |
75-05-8 | |
81°C | |
Liquid | |
1878 | |
0.36cp at 20°C | |
CC#N | |
41.05 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.