missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Honeywell

Product Code. 15675900
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
25 L
150 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Density 0.786g/mL at 25°C
Quantity 20 kg
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Beilstein 741857
Formula Weight 41.05g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'