missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
ACS Reagent,≥99.5%
92.78 € - 2129.00 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15625060
|
Honeywell-Riedel-de Haen
360457-500ML |
500 mL | Glass Bottle | ||||||
15695050
|
Honeywell-Riedel-de Haen
360457-1L |
1 L | Glass Bottle | ||||||
15605060
|
Honeywell-Riedel-de Haen
360457-2.5L |
2.5 L | Glass Bottle | ||||||
15685050
|
Honeywell-Riedel-de Haen
360457-10L |
10 L | Stainless Steel Can | ||||||
15615060
|
Honeywell-Riedel-de Haen
360457-20L |
20 L | Stainless Steel Can | ||||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
-48°C | |
0.786g/mL at 25°C | |
0.16 | |
973°F | |
Characteristic | |
0 to 0.005% | |
≤8μEQ/g | |
≤0.6μEQ/g | |
72.8mmHg (20°C) | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
Acetonitrile |
75-05-8 | |
81°C | |
2°C (35.6°F) | |
Conforms to structure | |
Liquid | |
C2H3N | |
CH3CN | |
MFCD00001878 | |
UN1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.5% |
Safety and Handling
P210-P261-P280-P305 + P351 + P338-P370 + P378-P403 + P235
missing translation for 'dotInformation' : 3/PGII
missing translation for 'rtecsNumber' : 200-835-2