missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15615060
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Density 0.786g/mL at 25°C
Boiling Point 81°C
Quantity 20 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Stainless Steel Can
Physical Form Liquid
Residue after Evaporation 0 to 0.005%
Titratable Acid ≤8μEQ/g
Linear Formula CH3CN
Titratable Base ≤0.6μEQ/g
Vapor Pressure 72.8mmHg (20°C)
UN Number UN1648
Beilstein 741857
Formula Weight 41.05g/mol
Percent Purity ≥99.5%
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'