missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)
315.70 € - 771.02 €
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15682550
|
Honeywell-Riedel-de Haen
24201-6X1L |
6 x 1 L | Poly Bottle |
315.70 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15652550
|
Honeywell-Riedel-de Haen
24201-4X2.5L-GL |
4 x 2.5 L | Glass Bottle |
451.36 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15642550
|
Honeywell-Riedel-de Haen
24201-4X2.5L |
4 x 2.5 L | Poly Bottle |
418.86 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15662550
|
Honeywell-Riedel-de Haen
24201-4X5L |
4 x 5 L | Poly Bottle |
771.02 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
-95°C | |
Colorless | |
55°C to 57°C | |
−17°C (1.4°F) | |
C3H6O | |
MFCD00008765 | |
UN1090 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99% (GC) | |
Acetone, Meets analytical specification of Ph. Eur.BP, NF |
67-64-1 | |
0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C | |
0.132 | |
Liquid | |
CH3COCH3 | |
184mmHg (20°C) | |
635680 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
puriss. |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319-H336
missing translation for 'dotInformation' : Class 3 - PG 2 - Acetone
missing translation for 'einecsNumber' : 200-662-2
missing translation for 'rtecsNumber' : AL3150000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title