missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid – Triethylamine Solution 1:1, Honeywell Fluka™
for HPLC, 2M:2M concentrate, in H2O
186.13 € - 668.91 €
Chemical Identifiers
CAS | 5204-74-0 |
---|---|
Molecular Formula | C8H19NO2 |
Molecular Weight (g/mol) | 161.245 |
MDL Number | MFCD00151725 |
InChI Key | AVBGNFCMKJOFIN-UHFFFAOYSA-N |
Synonym | triethylammonium acetate, teaa, acetic acid, compd. with n,n-diethylethanamine 1:1, acetic acid; n,n-diethylethanamine, ethanamine, n,n-diethyl-, acetate, triethylamine acetate, hoac et3n, acetic acid triethylamine, acetic acid; triethylamine, triethylammonium acetate solution |
PubChem CID | 165295 |
IUPAC Name | acetic acid;N,N-diethylethanamine |
SMILES | CCN(CC)CC.CC(=O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
5204-74-0 | |
161.245 | |
AVBGNFCMKJOFIN-UHFFFAOYSA-N | |
165295 | |
CCN(CC)CC.CC(=O)O |
C8H19NO2 | |
MFCD00151725 | |
triethylammonium acetate, teaa, acetic acid, compd. with n,n-diethylethanamine 1:1, acetic acid; n,n-diethylethanamine, ethanamine, n,n-diethyl-, acetate, triethylamine acetate, hoac et3n, acetic acid triethylamine, acetic acid; triethylamine, triethylammonium acetate solution | |
acetic acid;N,N-diethylethanamine |
Specifications
5204-74-0 | |
CH3COOH · N(CH2CH3)3 | |
NONH for all modes of transport | |
triethylammonium acetate, teaa, acetic acid, compd. with n,n-diethylethanamine 1:1, acetic acid; n,n-diethylethanamine, ethanamine, n,n-diethyl-, acetate, triethylamine acetate, hoac et3n, acetic acid triethylamine, acetic acid; triethylamine, triethylammonium acetate solution | |
CCN(CC)CC.CC(=O)O | |
161.245 | |
161.24g/mol | |
Acetic Acid – Triethylamine Solution 1:1 |
C8H19NO2 | |
MFCD00151725 | |
3688333 | |
AVBGNFCMKJOFIN-UHFFFAOYSA-N | |
acetic acid;N,N-diethylethanamine | |
165295 | |
1.010g/mL at 20°C |
Safety and Handling
P305 + P351 + P338
H315-H319-H335