missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ReagentPlus™, Honeywell

Product Code. 15677660
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethylic acid, methanecarboxylic acid, essigsaeure, vinegar acid, acetic acid, glacial, acide acetique, glacial, acetasol, ethanoic acid
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Quantity 1 L
Packaging Glass Bottle
Chemical Name or Material Acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'