missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial, USP, Macron Fine Chemicals™
Click to view available options
Quantity:
0.5 L
2.5 L
Packaging:
Clear bottle
Clear glass bottle
Description
- Meets the requirements necessary for use in pharmaceutical processing and prescription compounding
- Product is regulated by the Food and Drug Administration
- Manufactured under current Good Manufacturing Practices (cGMP)
Specifications
Specifications
pH | 2.5 (at 20°C) |
Melting Point | 16.6°C |
Boiling Point | 118°C |
Color | Colorless |
Appearance | Clear |
Assay Percent Range | 99.7 to 100.5% |
Linear Formula | CH3COOH |
Solubility Information | Fully miscible in water |
Formula Weight | 60.05g/mol |
Grade | USP |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'