missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, U.S.P. - F.C.C., J.T. Baker™
160.00 € - 281.00 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15578624
|
Avantor J.T.Baker
9522-02 |
500 mL | Glass Bottle |
216.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588624
|
Avantor J.T.Baker
9522-03 |
2.5 L | Glass Bottle |
160.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15598624
|
Avantor J.T.Baker
9522-05 |
2.5 L | Glass Bottle, Single ship |
281.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508634
|
Avantor J.T.Baker
9522-33 |
2.5 L | pvc coated glass bottle |
168.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
39.00°C | |
Acetic Acid, Glacial |
C2H4O2 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
FCC/USP | |
1L = 1.05kg |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.