missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals

Product Code. 13572310
Click to view available options
Quantity:
100 mL
500 mL
2 L
4 x 500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16.6°C
Boiling Point 118.1°C
Assay Percent Range 99.7+%
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water.
Formula Weight 60.05
Percent Purity ≥99.7%
Grade ACS
Density 1.049 g/mL
Quantity 2 L
Flash Point 40°C (104°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.3721
Chemical Name or Material Acetic acid, Glacial
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'