Learn More
Acetic acid, glacial, 99.7+% (metals basis), Thermo Scientific Chemicals
72.16 € - 280.40 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374717
|
Thermo Scientific Alfa Aesar
039745.AE |
100 mL |
72.16 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430241
|
Thermo Scientific Alfa Aesar
039745.AP |
500 mL |
280.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Good polar protic solvent, in its ester form it act as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeureShow More | |
CHEBI:15366 | |
CC(O)=O |
Specifications
2.5 | |
16.6°C | |
C2H4O2 | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
1.049 g/mL | |
Pungent | |
1.3721 | |
Acetic Acid, Glacial |
64-19-7 | |
118.1°C | |
MFCD00036152 | |
14,55 | |
Miscible with water,ethanol,acetone,toluene and immiscible in carbon disulphide. | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.9% | |
40°C (104°F) | |
Liquid | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
missing translation for 'einecsNumber' : 200-580-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.