missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, J.T. Baker™

Product Code. 10433191
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 64-19-7
Melting Point 16.6°C
Boiling Point 118°C
Assay Percent Range 99 to 100%
Molecular Formula C2H4O2
MDL Number MFCD00036152
UN Number 2789
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
Solubility Information Fully miscible in water
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight CH3COOH
Density 1.05g/cm3
Quantity 500 mL
Explosion Limit 4Vol% to 17Vol%
Flash Point 40°C
Ignition Point 485°C
Physical Form Liquid
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'