missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid concentrate, For 1L standard solution, 1.0 M CH3COOH (1N), Honeywell Fluka™
for 1L standard solution, 1.0 M CH3COOH (1N)
85.08 € - 313.07 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15605200
|
Honeywell-Fluka
38051-1EA |
1 Pc. |
85.08 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15615200
|
Honeywell-Fluka
38051-6X1EA |
6 x 1 Ea. | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
CH3CO2H | |
UN2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
40°C (104°F) | |
Acetic acid concentrate |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
Ampoule |
Safety and Handling
P280-P305 + P351 + P338-P310
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Acetic acid concentrate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title