missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid concentrate, For 1L standard solution, 0.1 M CH3COOH (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Min % | ≥80.0000%, ≥10.0000% |
CAS Max % | <90.0000%, <20.0000% |
Linear Formula | CH3CO2H |
UN Number | NONH for all modes of transport |
Beilstein | 506007 |
Formula Weight | 60.05 |
Quantity | 6 x 1 Ea. |
Packaging | Ampoule |
Physical Form | Concentrate |
Chemical Name or Material | Acetic acid concentrate, For 1L standard solution, 0.1 M CH3COOH (0.1N) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'