Learn More
α-Acetamidocinnamic acid, 98%, Thermo Scientific Chemicals
39.60 € - 109.55 €
Chemical Identifiers
CAS | 5469-45-4 |
---|---|
Molecular Formula | C11H11NO3 |
Molecular Weight (g/mol) | 205.21 |
InChI Key | XODAOBAZOQSFDS-YFHOEESVSA-N |
Synonym | z-2-acetamido-3-phenylacrylic acid, alpha-acetamidocinnamic acid, .alpha.-acetamidocinnamic acid, .alpha.-acetaminocinnamic acid, acetamidocinnamic acid, z-2-acetamido-3-phenylprop-2-enoic acid, 2-acetylamino-3-phenyl-2-propenoic acid, 2-acetamido-3-phenylprop-2-enoic acid, 2z-2-acetamido-3-phenylprop-2-enoic acid, pubchem8139 |
PubChem CID | 5370579 |
IUPAC Name | (Z)-2-acetamido-3-phenylprop-2-enoic acid |
SMILES | CC(=O)NC(=CC1=CC=CC=C1)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10149979
|
Thermo Scientific Acros
200260250 |
25 g | Glass bottle |
39.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11366095
|
Thermo Scientific Acros
200261000 |
100 g | Glass bottle |
109.55 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5469-45-4 | |
205.21 | |
z-2-acetamido-3-phenylacrylic acid, alpha-acetamidocinnamic acid, .alpha.-acetamidocinnamic acid, .alpha.-acetaminocinnamic acid, acetamidocinnamic acid, z-2-acetamido-3-phenylprop-2-enoic acid, 2-acetylamino-3-phenyl-2-propenoic acid, 2-acetamido-3-phenylprop-2-enoic acid, 2z-2-acetamido-3-phenylprop-2-enoic acid, pubchem8139 | |
(Z)-2-acetamido-3-phenylprop-2-enoic acid |
C11H11NO3 | |
XODAOBAZOQSFDS-YFHOEESVSA-N | |
5370579 | |
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O |
Specifications
5469-45-4 | |
97.5% min. (HPLC) | |
C6H5CH=C(NHCOCH3)CO2H | |
z-2-acetamido-3-phenylacrylic acid, alpha-acetamidocinnamic acid, .alpha.-acetamidocinnamic acid, .alpha.-acetaminocinnamic acid, acetamidocinnamic acid, z-2-acetamido-3-phenylprop-2-enoic acid, 2-acetylamino-3-phenyl-2-propenoic acid, 2-acetamido-3-phenylprop-2-enoic acid, 2z-2-acetamido-3-phenylprop-2-enoic acid, pubchem8139 | |
CC(=O)NC(=CC1=CC=CC=C1)C(=O)O | |
205.21 | |
205.21 | |
α-Acetamidocinnamic acid |
188.0°C to 190.0°C | |
C11H11NO3 | |
10683 | |
XODAOBAZOQSFDS-YFHOEESVSA-N | |
(Z)-2-acetamido-3-phenylprop-2-enoic acid | |
5370579 | |
0.98 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-795-6
- alpha-Acetamidocinnamic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only