Learn More
Acetaldehyde, 99.5%, extra pure, AcroSeal™, Thermo Scientific Chemicals
31.76 € - 73.97 €
Chemical Identifiers
CAS | 75-07-0 |
---|---|
Molecular Formula | C2H4O |
Molecular Weight (g/mol) | 44.04 |
MDL Number | MFCD00006991 |
InChI Key | IKHGUXGNUITLKF-UHFFFAOYSA-N |
Synonym | ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique |
PubChem CID | 177 |
ChEBI | CHEBI:15343 |
IUPAC Name | acetaldehyde |
SMILES | CC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10061304
|
Thermo Scientific Acros
427171000 |
100 mL | AcroSeal™ Glass bottle |
31.76 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15119153
|
Thermo Scientific Acros
427178000 |
800 mL | AcroSeal™ Glass bottle |
73.97 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-07-0 | |
44.04 | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
177 | |
acetaldehyde |
C2H4O | |
MFCD00006991 | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
CHEBI:15343 | |
CC=O |
Specifications
75-07-0 | |
100.0 | |
White | |
21.0°C | |
Authentic | |
C2H4O | |
MFCD00006991 | |
0.785 | |
Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone | |
CC=O | |
44.04 | |
CHEBI:15343 | |
99.5% | |
Solid |
99.4 | |
-123.0°C | |
0.7850g/mL | |
−27°C | |
99.4% min. (GC) | |
CH3CHO | |
15,4 | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
acetaldehyde | |
177 | |
44.04 | |
Extra Pure | |
Acetaldehyde, Extra Pure |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-836-8
- Acetaldehyde
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.