Learn More
Miele Professional™ Closed Plinth UG 30-60/60-85
Closed plinth UG 30-60/60-85 to raise the machine, 60 cm deep, 60 cm wide
Brand:  Miele Professional™ 10087350
Product Code. 15447724
  • 667.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless-steel plinth UG 30-60/60-85
  • For use on PG 8583 and PG 8593 Miele glass laboratory washers
  • Bolts on to machine
  • H 300mm, W 600mm, D 600mm