Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Brand:  Thermo Scientific™ RAC34567
 View more versions of this product
Product Code. 11336401
  • 892.44 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Factory-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Freezer Access Key
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series