missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Absorbance Test Plate
For use with any microplate reader with 340 nm absorbance measurement capability to evaluate optical systems at 340 nm
Brand: Agilent BioTek 7260551
850.25 EUR valid until 2024-12-31
Use promo code "21998" to get your promotional price.
This item is not returnable.
View return policy
Description
- Tests linearity, accuracy, repeatability and alignment
- Includes six filters for testing 340 nm at different optical densities
- With data sheet and instructions
Specifications
Absorbance Test Plate | |
Agilent BioTek Microplate Readers | |
for testing 340nm at different optical densities |