missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | ABHD6 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890035
|
Proteintech
20494-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880035
|
Proteintech
20494-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.Specifications
ABHD6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8R2Y0, Q9BV23 | |
57406, 66082 | |
ABHD6 Fusion Protein Ag13968 | |
Primary | |
-20°C | |
ABHD6 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ABHD6, Monoacylglycerol lipase ABHD6 | |
Abhd6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |