Learn More
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks
GREENER_CHOICE
Brand:  Thermo Scientific™ AB-0977
565.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11914794
  • 647.00 € / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks contain a lockable lid and silicone tube plugs for security during transport.
  • 96 individual 0.65 or 1.2mL tubes
  • Alphanumeric grid-referencing molded into the rack and lid
  • Temperature range -170°C to +120°C
  • Heavy-duty rack in a microplate format and in accordance with SBS footprint
  • Stackable with or without lid
  • Open bottom to allow full robotic access to micro-tubes—each tube is fully supported to withstand pressure applied by robotic systems
  • Sealing options include MULTISIP™ septum plugs, silicone tube plugs, storage plate cap strips
Ordering Alerts:
Supplied in packs of 10 racks, 96 tubes each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.2mL Grid-Referenced Cluster Tube Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks >