Learn More
Kimberly-Clark Professional™ KleenGuard™ A45 Breathable Liquid and Particle Protection Hooded Coveralls
Single-use, breathable hooded coveralls with storm flap for respirator. The ultimate for user comfort, made from high-performance fabric and film laminate. Anti-static and general protection against water-based or chemical splashes, liquids or dust particles. PPE Category III.
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material KleenGuard™
Color White
Disposable Single-use
View More Specs
Description
Description
  • Supplied in packs of 25
  • White, highly breathable, anti-static, silicone-free coveralls
  • High-performance fabric and film laminate construction for comfort, safety and strength.
  • Provides PPE Category III protection
  • Storm flap and hood designed for easier taping to a respirator
  • Provides anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • The ultimate combination of protection and user comfort in more critical areas.
  • Hood specially designed to fit a respirator, longer arms,
  • Full-length zip
Specifications
Specifications
General Purpose Coveralls
KleenGuard™
Single-use
Front Zipper
Water, Chemicals, Liquid, Particle
60 cm
Unisex
White
Hooded
Non-sterile
23.4 in.
25/Pk.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Kimberly-Clark Professional™ KleenGuard™ A45 Breathable Liquid and Particle Protection Hooded Coveralls