Learn More
Thermo Scientific™ ARCTIC A40 Refrigerated Circulator
A40 refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −40° to +200°C and 12L maximum bath volume.
Description
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ ARCTIC A40 Refrigerated Circulator