missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elma™ A10 Concentrated Cleaner
Providing powerful cleaning power for labware and medical instruments. Elma™ A10 Concentrated Cleaner is a mildly alkaline solution for dip and splash cleaning.
25.65 € - 169.00 €
Specifications
Product Type | Lab Clean A10 Cleaner |
---|---|
pH | 11.3 |
Certifications/Compliance | Annex VII A, EC 648/2004, UN 1719 |
Composition | Amphoteric surfactants 5 to 15%, non-ionic surfactants 5 to 15%, Phosphates <5%, Polycarboxylates |
Concentration | Ultrasonic unit 1 to 2%, dishwasher 0.5 to 1% |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11832681
|
Elma™
8000182 |
10 L |
169.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11822681
|
Elma™
8000181 |
2.5 L |
51.25 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10616295
|
Elma™
8000180 |
1 L |
25.65 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Suitable for laboratory instruments made of glass, porcelain, ceramics, plastics and metals (check aluminum, magnesium and light metal alloys before application)
- Removes emulsions, fat and grease, dust, glass grease, resinified residues, marker, labels, lime soaps and lime deposits
Specifications
Lab Clean A10 Cleaner | |
Annex VII A, EC 648/2004, UN 1719 | |
Ultrasonic unit 1 to 2%, dishwasher 0.5 to 1% | |
5°C to 30°C |
11.3 | |
Amphoteric surfactants 5 to 15%, non-ionic surfactants 5 to 15%, Phosphates <5%, Polycarboxylates | |
Bottle | |
For dip and splash cleaning of instruments of labs and workshops applications |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title