missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 9mm PE Snap Ring Seal for 8mm crimp vials, Transparent, Center hole, Assembled septum

Aluminum crimp seals with prefitted septa.

Brand:  Fisherbrand™ 09 15 0756

 View more versions of this product

Product Code. 11814830

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • Caps are easy to apply and easy to remove
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Snap-It caps eliminate the need for crimping or de-capping tools
  • Polyethylene caps are chemically inert and suitable for most chromatography applications
  • Provide a secure leak-resistant seal
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silicone/PTFE
Red, White, Transparent
Polypropylene
100
Snap Cap
4 mm
9 mm
GC, HPLC
1.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers