Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
For high-throughput screening in cell based test systems.
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Well Shape | Round Bottom (U-well) |
Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Free of detectable DNase, RNase, human DNA
- Prevents wicking
Specifications
Standard | |
Polypropylene | |
Round Bottom (U-well) |
8 x 12 | |
96 |
Safety and Handling
missing translation for 'shelfLife' : 5 Years