missing translation for 'onlineSavingsMsg'
Learn More

9-(Chloromethyl)anthracene, 98+%, Thermo Scientific Chemicals

35.98 € - 498.48 €

Chemical Identifiers

CAS 24463-19-2
Molecular Formula C15H11Cl
Molecular Weight (g/mol) 226.703
MDL Number MFCD00001263
InChI Key PCVRSXXPGXRVEZ-UHFFFAOYSA-N
Synonym 9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
PubChem CID 32385
IUPAC Name 9-(chloromethyl)anthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470094
View Documents
Thermo Scientific Alfa Aesar
A13579.03
1 g
35.98 €
1g
Please to purchase this item. Need a web account? Register with us today!
11480094
View Documents
Thermo Scientific Alfa Aesar
A13579.06
5 g
120.60 €
5g
Please to purchase this item. Need a web account? Register with us today!
11490094
View Documents
Thermo Scientific Alfa Aesar
A13579.14
25 g
498.48 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

24463-19-2
226.703
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
32385
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
C15H11Cl
MFCD00001263
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
9-(chloromethyl)anthracene

Specifications

24463-19-2
C15H11Cl
UN3261
9-chloromethyl anthracene, anthracene, 9-chloromethyl, 9-anthracenylmethyl chloride, ccris 7993, 9-chloromethyanthracene, pubchem14854, 9-chlormethyl-anthracen, acmc-209tez
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
226.703
226.71
9-(Chloromethyl)anthracene
138°C to 142°C
MFCD00001263
1873394
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-(chloromethyl)anthracene
32385
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'rtecsNumber' : CA9600000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.