Learn More
9,10-Anthraquinone, 98+%, Thermo Scientific Chemicals
82.11 €
Chemical Identifiers
CAS | 84-65-1 |
---|---|
Molecular Formula | C14H8O2 |
Molecular Weight (g/mol) | 208.22 |
MDL Number | MFCD00001188 |
InChI Key | RZVHIXYEVGDQDX-UHFFFAOYSA-N |
Synonym | anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone |
PubChem CID | 6780 |
ChEBI | CHEBI:40448 |
IUPAC Name | anthracene-9,10-dione |
SMILES | O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400693
|
Thermo Scientific Alfa Aesar
A12735.30 |
250 g |
82.11 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490683
|
Thermo Scientific Alfa Aesar
|
1000 g |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Anthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.
Solubility
Insoluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
84-65-1 | |
208.22 | |
RZVHIXYEVGDQDX-UHFFFAOYSA-N | |
6780 | |
anthracene-9,10-dione |
C14H8O2 | |
MFCD00001188 | |
anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone | |
CHEBI:40448 | |
O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12 |
Specifications
84-65-1 | |
1.438 | |
185°C (365°F) | |
C14H8O2 | |
390030 | |
anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone | |
RZVHIXYEVGDQDX-UHFFFAOYSA-N | |
anthracene-9,10-dione | |
6780 | |
208.22 | |
9,10-Anthraquinone |
283°C to 287°C | |
377°C | |
Odorless | |
MFCD00001188 | |
14,687 | |
Insoluble in water. | |
O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12 | |
208.22 | |
CHEBI:40448 | |
≥98% |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
missing translation for 'einecsNumber' : 201-549-0
missing translation for 'rtecsNumber' : CB4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.