missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyjulolidine, 95%, Thermo Scientific Chemicals

Product Code. 15407017
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63468-83-7
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.29
MDL Number MFCD00037096
InChI Key ZSKPEBROOQBZNW-UHFFFAOYSA-N
Synonym 8-methoxyjulolidine, 8-methoxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, enamine_005788, 8-methoxy-1-azatricyclo 7.3.1.0?, 1 3 trideca-5,7,9 13-triene
PubChem CID 2829840
IUPAC Name 6-methoxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
SMILES COC1=C2CCCN3CCCC(C=C1)=C23

Specifications

Quantity 1 g
Formula Weight 203.28
Percent Purity 95%
Chemical Name or Material 8-Methoxyjulolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'