missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-methoxycoumarin, 97%, Thermo Scientific™

63.00 € - 234.00 €

Chemical Identifiers

CAS 19492-03-6
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00075652
InChI Key KIGCGZUAVODHMD-UHFFFAOYSA-N
PubChem CID 146487
IUPAC Name 8-hydroxy-7-methoxychromen-2-one
SMILES COC1=C(C2=C(C=C1)C=CC(=O)O2)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383277
View Documents
Thermo Scientific Alfa Aesar
L13841.MC
100 mg
63.00 €
100mg
Please to purchase this item. Need a web account? Register with us today!
11373277
View Documents
Thermo Scientific Alfa Aesar
L13841.ME
500 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

19492-03-6
192.17
KIGCGZUAVODHMD-UHFFFAOYSA-N
8-hydroxy-7-methoxychromen-2-one
C10H8O4
MFCD00075652
146487
COC1=C(C2=C(C=C1)C=CC(=O)O2)O

Specifications

19492-03-6
Odorless
MFCD00075652
8-hydroxy-7-methoxycoumarin, daphnetin-7-methylether, 8-hydroxy-7-methoxy-2h-chromen-2-one, 8-hydroxy-7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 8-hydroxy-7-methoxy, 2h-1-benzopyran-2-one,8-hydroxy-7-methoxy, acmc-20alwe, daphnetin 7-methylether, daphnetin 7-methyl ether, daphnetin-7-methyl ether
COC1=C(C2=C(C=C1)C=CC(=O)O2)O
192.17
192.17
8-Hydroxy-7-methoxycoumarin
175°C to 177°C
C10H8O4
168393
KIGCGZUAVODHMD-UHFFFAOYSA-N
8-hydroxy-7-methoxychromen-2-one
146487
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.